ADVH49B83360 AChemBlock

(S)-4-(1-aminoethyl)aniline dihydrochloride

  • Cas Number : 255060-76-5
  • Molecular Weight : 209.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/s-4-1-aminoethyl-aniline-dihydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87829
Availability details for
Quantity
  • Availability
  • Add to Cart