ADVH49B83366 AChemBlock

5-methyl-1,2,3,4-tetrahydronaphthalene

  • Cas Number : 2809-64-5
  • Molecular Weight : 146.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/5-methyl-1-2-3-4-tetrahydronaphthalene.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87835
Availability details for
Quantity
  • Availability
  • Add to Cart