ADVH49B83369 AChemBlock

(2-bromo-4-fluorophenyl)methanamine hydrochloride

  • Cas Number : 289038-14-8
  • Molecular Weight : 240.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/2-bromo-4-fluorophenyl-methanamine-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87838
Availability details for
Quantity
  • Availability
  • Add to Cart