ADVH49B8336E AChemBlock

(R)-2-(4-fluorophenyl)pyrrolidine

  • Cas Number : 298690-89-8
  • Molecular Weight : 165.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/r-2-4-fluorophenyl-pyrrolidine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87843
Availability details for
Quantity
  • Availability
  • Add to Cart