ADVH49B83375 AChemBlock

2,2,2-trifluoro-1-(o-tolyl)ethan-1-amine hydrochloride

  • Cas Number : 315-43-5
  • Molecular Weight : 225.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/2-2-2-trifluoro-1-o-tolyl-ethan-1-amine-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87850
Availability details for
Quantity
  • Availability
  • Add to Cart