ADVH49B83377 AChemBlock

(S)-3-(1-aminoethyl)aniline

  • Cas Number : 317830-29-8
  • Molecular Weight : 136.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/s-3-1-aminoethyl-aniline.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87852
Availability details for
Quantity
  • Availability
  • Add to Cart