ADVH49B83378 AChemBlock

5-bromochromane

  • Cas Number : 319478-14-3
  • Molecular Weight : 213.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/5-bromochromane.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87853
Availability details for
Quantity
  • Availability
  • Add to Cart