ADVH49B8337A AChemBlock

(R)-1-(3,4-difluorophenyl)ethan-1-amine

  • Cas Number : 321318-15-4
  • Molecular Weight : 157.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/r-1-3-4-difluorophenyl-ethan-1-amine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87855
Availability details for
Quantity
  • Availability
  • Add to Cart