ADVH49B8337D AChemBlock

4,4'-carbonyldibenzonitrile

  • Cas Number : 32446-66-5
  • Molecular Weight : 232.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/4-4-carbonyldibenzonitrile.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87858
Availability details for
Quantity
  • Availability
  • Add to Cart