ADVH49B8337F AChemBlock

2,2,2-trifluoro-1-(naphthalen-1-yl)ethan-1-amine

  • Cas Number : 326594-18-7
  • Molecular Weight : 225.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/2-2-2-trifluoro-1-naphthalen-1-yl-ethan-1-amine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87860
Availability details for
Quantity
  • Availability
  • Add to Cart