ADVH49B83380 AChemBlock

chromane-8-carbaldehyde

  • Cas Number : 327183-32-4
  • Molecular Weight : 162.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/chromane-8-carbaldehyde.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87861
Availability details for
Quantity
  • Availability
  • Add to Cart