ADVH49B83385 AChemBlock

2-(4-methoxyphenyl)piperidine hydrochloride

  • Cas Number : 341526-79-2
  • Molecular Weight : 227.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/2-4-methoxyphenyl-piperidine-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87866
Availability details for
Quantity
  • Availability
  • Add to Cart