ADVH49B83387 AChemBlock

methyl (S)-2-amino-3-(3-hydroxyphenyl)propanoate hydrochloride

  • Cas Number : 34260-72-5
  • Molecular Weight : 231.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/methyl-s-2-amino-3-3-hydroxyphenyl-propanoate-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87868
Availability details for
Quantity
  • Availability
  • Add to Cart