ADVH49B8338C AChemBlock

8-methoxynaphthalen-1-ol

  • Cas Number : 3588-75-8
  • Molecular Weight : 174.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/8-methoxynaphthalen-1-ol.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87873
Availability details for
Quantity
  • Availability
  • Add to Cart