ADVH49B8338F AChemBlock

8-methylchromane

  • Cas Number : 3722-72-3
  • Molecular Weight : 148.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/8-methylchromane.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87876
Availability details for
Quantity
  • Availability
  • Add to Cart