ADVH49B83391 AChemBlock

1-(2,3-dihydrobenzofuran-6-yl)ethan-1-one

  • Cas Number : 374706-07-7
  • Molecular Weight : 162.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/1-2-3-dihydrobenzofuran-6-yl-ethan-1-one.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87878
Availability details for
Quantity
  • Availability
  • Add to Cart