ADVH49B83393 AChemBlock

4-methoxybenzo[b]thiophene

  • Cas Number : 3781-90-6
  • Molecular Weight : 164.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/4-methoxybenzo-b-thiophene.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87880
Availability details for
Quantity
  • Availability
  • Add to Cart