ADVH49B83395 AChemBlock

2-(2,6-dichlorophenyl)pyrrolidine

  • Cas Number : 383127-39-7
  • Molecular Weight : 216.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/2-2-6-dichlorophenyl-pyrrolidine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87882
Availability details for
Quantity
  • Availability
  • Add to Cart