ADVH49B8339C AChemBlock

methyl 4-oxochromane-7-carboxylate

  • Cas Number : 41118-21-2
  • Molecular Weight : 206.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/methyl-4-oxochromane-7-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87889
Availability details for
Quantity
  • Availability
  • Add to Cart