ADVH49B8339F AChemBlock

3-methyl-1-phenylbutan-1-amine

  • Cas Number : 42290-97-1
  • Molecular Weight : 163.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/3-methyl-1-phenylbutan-1-amine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87892
Availability details for
Quantity
  • Availability
  • Add to Cart