ADVH49B833A2 AChemBlock

1-(5-methylpyridin-3-yl)ethan-1-one

  • Cas Number : 42972-46-3
  • Molecular Weight : 135.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/1-5-methylpyridin-3-yl-ethan-1-one.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87895
Availability details for
Quantity
  • Availability
  • Add to Cart