ADVH49B833A4 AChemBlock

(S)-1-(3-fluorophenyl)ethan-1-amine

  • Cas Number : 444643-09-8
  • Molecular Weight : 139.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/s-1-3-fluorophenyl-ethan-1-amine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87897
Availability details for
Quantity
  • Availability
  • Add to Cart