ADVH49B833A5 AChemBlock

3-(difluoromethyl)-4-fluoroaniline

  • Cas Number : 445303-96-8
  • Molecular Weight : 161.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/3-difluoromethyl-4-fluoroaniline.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87898
Availability details for
Quantity
  • Availability
  • Add to Cart