ADVH49B833A6 AChemBlock

2,3,5,6-tetrahydro-7H-indeno[5,6-b]furan-7-one

  • Cas Number : 448964-30-5
  • Molecular Weight : 174.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/2-3-5-6-tetrahydro-7h-indeno-5-6-b-furan-7-one.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87899
Availability details for
Quantity
  • Availability
  • Add to Cart