ADVH49B833AA AChemBlock

1-(3-(trifluoromethyl)phenyl)propan-1-amine

  • Cas Number : 473732-59-1
  • Molecular Weight : 203.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/1-3-trifluoromethyl-phenyl-propan-1-amine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87903
Availability details for
Quantity
  • Availability
  • Add to Cart