ADVH49B833AD AChemBlock

5-methoxyisobenzofuran-1(3H)-one

  • Cas Number : 4741-62-2
  • Molecular Weight : 164.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/5-methoxyisobenzofuran-1-3h-one.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87906
Availability details for
Quantity
  • Availability
  • Add to Cart