ADVH49B833AF AChemBlock

benzofuran-4-ol

  • Cas Number : 480-97-7
  • Molecular Weight : 134.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/benzofuran-4-ol.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87908
Availability details for
Quantity
  • Availability
  • Add to Cart