ADVH49B833B0 AChemBlock

(S)-3-(pyrrolidin-2-yl)pyridine

  • Cas Number : 494-97-3
  • Molecular Weight : 148.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/s-3-pyrrolidin-2-yl-pyridine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87909
Availability details for
Quantity
  • Availability
  • Add to Cart