ADVH49B833C6 AChemBlock

chroman-7-ol

  • Cas Number : 57052-72-9
  • Molecular Weight : 150.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/chroman-7-ol.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87931
Availability details for
Quantity
  • Availability
  • Add to Cart