ADVH49B833C7 AChemBlock

6-fluoro-1,2,3,4-tetrahydronaphthalen-1-ol

  • Cas Number : 58485-69-1
  • Molecular Weight : 166.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/6-fluoro-1-2-3-4-tetrahydronaphthalen-1-ol.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87932
Availability details for
Quantity
  • Availability
  • Add to Cart