ADVH49B833CA AChemBlock

6-bromo-2-methoxynicotinaldehyde

  • Cas Number : 58819-88-8
  • Molecular Weight : 216.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/6-bromo-2-methoxynicotinaldehyde.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87935
Availability details for
Quantity
  • Availability
  • Add to Cart