ADVH49B833CB AChemBlock

(S)-2-phenylpyrrolidine

  • Cas Number : 59347-91-0
  • Molecular Weight : 147.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/s-2-phenylpyrrolidine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87936
Availability details for
Quantity
  • Availability
  • Add to Cart