ADVH49B833D1 AChemBlock

5-methyl-1,3,4-thiadiazole-2-carbaldehyde

  • Cas Number : 61018-49-3
  • Molecular Weight : 128.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/5-methyl-1-3-4-thiadiazole-2-carbaldehyde.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87942
Availability details for
Quantity
  • Availability
  • Add to Cart