ADVH49B833D2 AChemBlock

4-bromo-2,3-dihydroxybenzoic acid

  • Cas Number : 61203-52-9
  • Molecular Weight : 233.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/4-bromo-2-3-dihydroxybenzoic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87943
Availability details for
Quantity
  • Availability
  • Add to Cart