ADVH49B833D4 AChemBlock

6-methoxy-7-methyl-3,4-dihydronaphthalen-1(2H)-one

  • Cas Number : 61495-10-1
  • Molecular Weight : 190.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/6-methoxy-7-methyl-3-4-dihydronaphthalen-1-2h-one.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87945
Availability details for
Quantity
  • Availability
  • Add to Cart