ADVH49B833D5 AChemBlock

3-(dimethylamino)benzaldehyde

  • Cas Number : 619-22-7
  • Molecular Weight : 149.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/3-dimethylamino-benzaldehyde.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87946
Availability details for
Quantity
  • Availability
  • Add to Cart