ADVH49B833D6 AChemBlock

(S)-1-(3-bromophenyl)propan-1-amine hydrochloride

  • Cas Number : 623143-34-0
  • Molecular Weight : 250.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/s-1-3-bromophenyl-propan-1-amine-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87947
Availability details for
Quantity
  • Availability
  • Add to Cart