ADVH49B833DC AChemBlock

2-(4-methoxyphenyl)piperidine

  • Cas Number : 63359-20-6
  • Molecular Weight : 191.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/2-4-methoxyphenyl-piperidine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87953
Availability details for
Quantity
  • Availability
  • Add to Cart