ADVH49B833E0 AChemBlock

methyl 4-bromo-5-formylthiophene-2-carboxylate

  • Cas Number : 648412-59-3
  • Molecular Weight : 249.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/methyl-4-bromo-5-formylthiophene-2-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87957
Availability details for
Quantity
  • Availability
  • Add to Cart