ADVH49B833E1 AChemBlock

(S)-2-amino-2-(naphthalen-1-yl)acetic acid hydrochloride

  • Cas Number : 649554-52-9
  • Molecular Weight : 237.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/s-2-amino-2-naphthalen-1-yl-acetic-acid-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87958
Availability details for
Quantity
  • Availability
  • Add to Cart