ADVH49B833E3 AChemBlock

2,2,2-trifluoro-1-(furan-2-yl)ethan-1-amine hydrochloride

  • Cas Number : 65686-91-1
  • Molecular Weight : 201.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/2-2-2-trifluoro-1-furan-2-yl-ethan-1-amine-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87960
Availability details for
Quantity
  • Availability
  • Add to Cart