ADVH49B833EB AChemBlock

ethyl 2-oxo-5,6,7,8-tetrahydro-2H-chromene-3-carboxylate

  • Cas Number : 66979-47-3
  • Molecular Weight : 222.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/ethyl-2-oxo-5-6-7-8-tetrahydro-2h-chromene-3-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87968
Availability details for
Quantity
  • Availability
  • Add to Cart