ADVH49B833F0 AChemBlock

6-methoxychroman-4-amine hydrochloride

  • Cas Number : 67858-19-9
  • Molecular Weight : 215.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/6-methoxychroman-4-amine-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87973
Availability details for
Quantity
  • Availability
  • Add to Cart