ADVH49B833F4 AChemBlock

methyl 1-oxo-2,3-dihydro-1H-indene-5-carboxylate

  • Cas Number : 68634-02-6
  • Molecular Weight : 190.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/methyl-1-oxo-2-3-dihydro-1h-indene-5-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87977
Availability details for
Quantity
  • Availability
  • Add to Cart