ADVH49B833FD AChemBlock

5-fluoroisobenzofuran-1(3H)-one

  • Cas Number : 700-85-6
  • Molecular Weight : 152.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/5-fluoroisobenzofuran-1-3h-one.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87986
Availability details for
Quantity
  • Availability
  • Add to Cart