ADVH49B833FE AChemBlock

7-(trifluoromethyl)chroman-4-amine

  • Cas Number : 704208-25-3
  • Molecular Weight : 217.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/7-trifluoromethyl-chroman-4-amine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87987
Availability details for
Quantity
  • Availability
  • Add to Cart