ADVH49B83406 AChemBlock

7-hydroxychroman-4-one

  • Cas Number : 76240-27-2
  • Molecular Weight : 164.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/7-hydroxychroman-4-one.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87995
Availability details for
Quantity
  • Availability
  • Add to Cart