ADVH49B83407 AChemBlock

(S)-4-(1-amino-2-hydroxyethyl)phenol hydrochloride

  • Cas Number : 763139-04-4
  • Molecular Weight : 189.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/s-4-1-amino-2-hydroxyethyl-phenol-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87996
Availability details for
Quantity
  • Availability
  • Add to Cart