ADVH49B83409 AChemBlock

1-mesitylethan-1-amine

  • Cas Number : 76808-14-5
  • Molecular Weight : 163.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/1-mesitylethan-1-amine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S87998
Availability details for
Quantity
  • Availability
  • Add to Cart