ADVH49B8340B AChemBlock

(R)-1-(5-methylfuran-2-yl)propan-1-amine hydrochloride

  • Cas Number : 779340-50-0
  • Molecular Weight : 175.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/r-1-5-methylfuran-2-yl-propan-1-amine-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S88000
Availability details for
Quantity
  • Availability
  • Add to Cart